Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PF-04708671 98%

Supplier: AMBEED, INC
Warning

A130606-10MG A130606-1MG A130606-50MG A130606-5MG A130606-100MG
77149-582EA 61 USD InStock
77149-582 77149-584 77149-586 77149-588 77685-137
PF-04708671 98%
PF-04708671
Precaución: for Research Use Only


Formula: C₁₉H₂₁F₃N₆
MDL Number: MFCD18086922
CAS Number: 1255517-76-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty