Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-Methoxypiperidine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A133907-1G A133907-250MG A133907-5G
77149-752EA 45 USD InStock
77149-752 77149-754 77149-756
(R)-3-Methoxypiperidine hydrochloride 98%
(R)-3-Methoxypiperidine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₃NO·ClH
MW: 151.634 g/mol
MDL Number: MFCD17015899
CAS Number: 688809-95-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty