Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-6-methoxypicolinic acid 95%

Supplier: AMBEED, INC
Warning

A134037-100MG A134037-1G A134037-250MG A134037-50MG
77149-758EA 95 USD InStock
77149-758 77149-760 77149-762 77149-764
5-Fluoro-6-methoxypicolinic acid 95%
5-Fluoro-6-methoxypicolinic acid
Precaución: for Research Use Only


Formula: C₇H₆FNO₃
MDL Number: MFCD13185872
CAS Number: 1211534-53-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty