Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Toceranib 98%

Supplier: AMBEED, INC
Warning

A135686-10MG A135686-1MG A135686-250MG A135686-25MG A135686-50MG A135686-5MG
77149-810EA 9 USD InStock
77149-810 77149-812 77149-814 77149-816 77149-818 77149-820
Toceranib 98%
Toceranib
Precaución: for Research Use Only


Formula: C₂₂H₂₅FN₄O₂
MDL Number: MFCD16038046
CAS Number: 356068-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty