Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

14-Methyl-8,14-dihydroindolo[2',3':3,4]pyrido[2,1-b]quinazolin-5(7H)-one hydrochloride 95%

Supplier: AMBEED, INC
Warning

A137270-1MG A137270-5MG
77149-900EA 24 USD InStock
77149-900 77149-902
14-Methyl-8,14-dihydroindolo[2',3':3,4]pyrido[2,1-b]quinazolin-5(7H)-one hydrochloride 95%
14-Methyl-8,14-dihydroindolo[2',3':3,4]pyrido[2,1-b]quinazolin-5(7H)-one hydrochloride
Precaución: for Research Use Only


Formula: C₁₉H₁₆ClN₃O
MDL Number: MFCD32061978
CAS Number: 111664-82-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty