Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-3-iodophenyltrifluoroborate 98+%

Supplier: AMBEED, INC
Warning

A138945-1G A138945-250MG A138945-5G
77149-964EA 56 USD InStock
77149-964 77149-966 77149-968
Potassium-3-iodophenyltrifluoroborate 98+%
Potassium 3-iodophenyltrifluoroborate
Precaución: for Research Use Only


Formula: C₆H₄BIF₃K
MDL Number: MFCD09800739
CAS Number: 1189097-36-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty