Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-3-tritylamino-4-[(2-N-bocamino)ethylaminoacarbonyl]amino-2H-pyrazole 95%

Supplier: AMBEED, INC
Warning

A140701-1G A140701-250MG A140701-5G
77150-042EA 24 USD InStock
77150-042 77150-044 77150-046
2-Methyl-3-tritylamino-4-[(2-N-bocamino)ethylaminoacarbonyl]amino-2H-pyrazole 95%
2-Methyl-3-tritylamino-4-[(2-N-bocamino)ethylaminoacarbonyl]amino-2H-pyrazole
Precaución: for Research Use Only


Formula: C₃₁H₃₆N₆O₃
MDL Number: MFCD24848694
CAS Number: 689293-69-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty