Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Cbz-glycine(Z-Gly-OH) 98%

Supplier: AMBEED, INC
Warning

A141974-100G A141974-10G A141974-25G A141974-500G A141974-1000G
77150-112EA 36 USD InStock
77150-112 77150-114 77150-116 77150-118 77685-209
N-Cbz-glycine(Z-Gly-OH) 98%
N-Cbz-glycine(Z-Gly-OH)
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00002691
CAS Number: 1138-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty