Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mycophenolate mofetil 98+%

Supplier: AMBEED, INC
Warning

A143691-10G A143691-1G A143691-250MG A143691-25G A143691-50MG A143691-5G A143691-100G
77150-160EA 41 USD InStock
77150-160 77150-162 77150-164 77150-166 77150-168 77150-170 77685-216
Mycophenolate mofetil 98+%
Mycophenolate mofetil
Precaución: for Research Use Only


Formula: C₂₃H₃₁NO₇
MW: 433.50 g/mol
MDL Number: MFCD00867568
CAS Number: 128794-94-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty