Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MK-2206 dihydrochloride 98%

Supplier: AMBEED, INC
Warning

A145592-10MG A145592-1MG A145592-1G
77150-244EA 43 USD InStock
77150-244 77150-246 77685-234
MK-2206 dihydrochloride 98%
MK-2206 dihydrochloride
Precaución: for Research Use Only


Formula: C₂₅H₂₁N₅O·2HCl
MDL Number: MFCD14584463
CAS Number: 1032350-13-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty