Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nistose trihydrate 95%

Supplier: AMBEED, INC
Warning

A148399-10MG A148399-1MG
77150-308EA 38 USD InStock
77150-308 77150-310
Nistose trihydrate 95%
Nistose trihydrate
Precaución: for Research Use Only


Formula: C₂₄H₄₂O₂₁
MW: 666.58 g/mol
MDL Number: MFCD00191672
CAS Number: 13133-07-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty