Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,4S,4aR,9aS)-rel-4a-Methyl-1,4,4a,9a-tetrahydro-1,4-methanoanthracene-9,10-dione 97%

Supplier: AMBEED, INC
Warning

A149615-100G A149615-10G A149615-1G A149615-250MG A149615-25G A149615-5G
77150-340EA 264.01 USD InStock
77150-340 77150-342 77150-344 77150-346 77150-348 77150-350
(1R,4S,4aR,9aS)-rel-4a-Methyl-1,4,4a,9a-tetrahydro-1,4-methanoanthracene-9,10-dione 97%
(1R,4S,4aR,9aS)-rel-4a-Methyl-1,4,4a,9a-tetrahydro-1,4-methanoanthracene-9,10-dione
Precaución: for Research Use Only


Formula: C₁₆H₁₄O₂
MDL Number: MFCD29044886
CAS Number: 97804-50-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty