Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[5-(Trifluoromethyl)pyridin-2-yl]acetonitrile 95%

Supplier: AMBEED, INC
Warning

A154001-100MG A154001-1G A154001-250MG A154001-5G
77150-530EA 13 USD InStock
77150-530 77150-532 77150-534 77150-536
[5-(Trifluoromethyl)pyridin-2-yl]acetonitrile 95%
[5-(Trifluoromethyl)pyridin-2-yl]acetonitrile
Precaución: for Research Use Only


Formula: C₈H₅F₃N₂
MDL Number: MFCD08690162
CAS Number: 939793-18-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty