Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GSK2606414 ≥98%

Supplier: AMBEED, INC
Warning

A154425-1MG A154425-2MG A154425-5MG
77150-572EA 13 USD InStock
77150-572 77150-574 77150-576
GSK2606414 ≥98%
GSK2606414
Precaución: for Research Use Only


Formula: C₂₄H₂₀F₃N₅O
MW: 451.4 g/mol
MDL Number: MFCD25976926
CAS Number: 1337531-36-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty