Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Levulinic acid 97%

Supplier: AMBEED, INC
Warning

A155719-1000G A155719-100G A155719-25G A155719-500G
77150-650EA 39 USD InStock
77150-650 77150-652 77150-654 77150-656
Levulinic acid 97%
Levulinic acid
Precaución: for Research Use Only


Formula: C₅H₈O₃
MDL Number: MFCD00002796
CAS Number: 123-76-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty