Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Esomeprazole magnesium trihydrate 98%

Supplier: AMBEED, INC
Warning

A157451-100MG A157451-25G A157451-50MG A157451-5G A157451-100G
77150-716EA 7 USD InStock
77150-716 77150-718 77150-720 77150-722 77685-298
Esomeprazole magnesium trihydrate 98%
Esomeprazole magnesium trihydrate
Precaución: for Research Use Only


Formula: C₃₄H₄₂MgN₆O₉S₂
MW: 767.16 g/mol
MDL Number: MFCD07698573
CAS Number: 217087-09-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty