Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Aminomethyl)cyclopropanecarbonitrile hydrochloride 95%

Supplier: AMBEED, INC
Warning

A160068-100MG A160068-1G A160068-250MG A160068-5G
77150-820EA 25 USD InStock
77150-820 77150-822 77150-824 77150-826
1-(Aminomethyl)cyclopropanecarbonitrile hydrochloride 95%
1-(Aminomethyl)cyclopropanecarbonitrile hydrochloride
Precaución: for Research Use Only


Formula: C₅H₉ClN₂
MDL Number: MFCD19382420
CAS Number: 1205544-78-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty