Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ML-323 98%

Supplier: AMBEED, INC
Warning

A160586-10MG A160586-1MG A160586-50MG A160586-100MG A160586-250MG
77150-868EA 85 USD InStock
77150-868 77150-870 77150-872 77685-315 77685-316
ML-323 98%
ML-323


Formula: C₂₃H₂₄N₆
MDL Number: MFCD28099817
CAS Number: 1572414-83-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty