Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-7762 ≥98%

Supplier: AMBEED, INC
Warning

A165759-10MG A165759-1MG A165759-5MG
77151-070EA 53 USD InStock
77151-070 77151-072 77151-074
AZD-7762 ≥98%
AZD-7762
Precaución: for Research Use Only


Formula: C₁₇H₁₉FN₄O₂S
MW: 362.42 g/mol
MDL Number: MFCD12406007
CAS Number: 860352-01-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty