Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4,4-difluorocyclohexanecarboxylate 98%

Supplier: AMBEED, INC
Warning

A170009-1G A170009-250MG A170009-5G
77151-232EA 24 USD InStock
77151-232 77151-234 77151-236
Methyl-4,4-difluorocyclohexanecarboxylate 98%
Methyl-4,4-difluorocyclohexanecarboxylate
Precaución: for Research Use Only


Formula: C₈H₁₂F₂O₂
MDL Number: MFCD11040454
CAS Number: 121629-14-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty