Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-Orn(fmoc)-OH 95%

Supplier: AMBEED, INC
Warning

A172205-100MG A172205-1G A172205-250MG A172205-25G A172205-5G
77151-308EA 8 USD InStock
77151-308 77151-310 77151-312 77685-382 77685-383
H-Orn(fmoc)-OH 95%
H-Orn(fmoc)-OH


Formula: C₂₀H₂₂N₂O₄
MDL Number: MFCD00237208
CAS Number: 147071-84-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty