Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-4-tert-Butyl 1-methyl 2-aminosuccinate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A174408-100MG A174408-1G A174408-250MG
77151-416EA 7 USD InStock
77151-416 77151-418 77151-420
(R)-4-tert-Butyl 1-methyl 2-aminosuccinate hydrochloride 95%
(R)-4-tert-Butyl 1-methyl 2-aminosuccinate hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₈ClNO₄
MW: 239.69 g/mol
MDL Number: MFCD04973240
CAS Number: 63329-02-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty