Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-(((trifluoromethyl)sulfonyl)oxy)-5,6-dihydropyridine-1(2H)-carboxylate 97%

Supplier: AMBEED, INC
Danger

A174808-250MG A174808-25G
77151-448EA 7 USD InStock
77151-448 77685-392
tert-Butyl-4-(((trifluoromethyl)sulfonyl)oxy)-5,6-dihydropyridine-1(2H)-carboxylate 97%
tert-Butyl-4-(((trifluoromethyl)sulfonyl)oxy)-5,6-dihydropyridine-1(2H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₆F₃NO₅S
MW: 331.3087 g/mol
MDL Number: MFCD09997858
CAS Number: 138647-49-1
UN: 1760
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty