Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Bis(trimethylstannyl)thieno[3,2-b]thiophene 98%

Supplier: AMBEED, INC
Danger

A175967-100MG A175967-1G A175967-250MG
77151-488EA 28 USD InStock
77151-488 77151-490 77151-492
2,5-Bis(trimethylstannyl)thieno[3,2-b]thiophene 98%
2,5-Bis(trimethylstannyl)thieno[3,2-b]thiophene
Precaución: for Research Use Only


Formula: C₁₂H₂₀S₂Sn₂
MDL Number: MFCD22200503
CAS Number: 469912-82-1
UN: 3146
ADR: 6.1,Ⅰ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty