Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Z)-5H-Cyclopenta[b]pyridin-7(6H)-one O-methyl oxime 98%

Supplier: AMBEED, INC
Danger

A176492-1G A176492-250MG
77151-504EA 936 USD InStock
77151-504 77151-506
(Z)-5H-Cyclopenta[b]pyridin-7(6H)-one O-methyl oxime 98%
(Z)-5H-Cyclopenta[b]pyridin-7(6H)-one O-methyl oxime
Precaución: for Research Use Only


Formula: C₉H₁₀N₂O
MDL Number: MFCD28975314
CAS Number: 1071727-78-0
UN: 1325
ADR: 4.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty