Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4R)-2,2-Dioxido-4-methyl-1,2,3-oxathiazolidine, N-BOC protected 98%

Supplier: AMBEED, INC
Warning

A177317-100G A177317-10G A177317-1G A177317-250MG A177317-25G A177317-5G
77151-518EA 450.01 USD InStock
77151-518 77151-520 77151-522 77151-524 77151-526 77151-528
(4R)-2,2-Dioxido-4-methyl-1,2,3-oxathiazolidine, N-BOC protected 98%
(4R)-2,2-Dioxido-4-methyl-1,2,3-oxathiazolidine, N-BOC protected
Precaución: for Research Use Only


Formula: C₈H₁₅NO₅S
MDL Number: MFCD09864994
CAS Number: 454248-53-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty