Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium (4-iodophenyl)trifluoroborate 98%

Supplier: AMBEED, INC
Warning

A177439-1G A177439-250MG A177439-5G
77151-530EA 46 USD InStock
77151-530 77151-532 77151-534
Potassium (4-iodophenyl)trifluoroborate 98%
Potassium (4-iodophenyl)trifluoroborate
Precaución: for Research Use Only


Formula: C₆H₄BF₃IK
MDL Number: MFCD09800738
CAS Number: 912350-00-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty