Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ezatiostat 98%

Supplier: AMBEED, INC
Warning

A178200-10MG A178200-1MG A178200-50MG A178200-5MG
77151-544EA 128 USD InStock
77151-544 77151-546 77151-548 77151-550
Ezatiostat 98%
Ezatiostat
Precaución: for Research Use Only


Formula: C₂₇H₃₅N₃O₆S
MW: 529.65 g/mol
MDL Number: MFCD27982957
CAS Number: 168682-53-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty