Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Amino-1-methylcyclohexanol 98%

Supplier: AMBEED, INC
Danger

A180629-100MG A180629-250MG A180629-5G
77151-582EA 87 USD InStock
77151-582 77151-584 77151-586
trans-4-Amino-1-methylcyclohexanol 98%
trans-4-Amino-1-methylcyclohexanol
Precaución: for Research Use Only


Formula: C₇H₁₅NO
MDL Number: MFCD18632623
CAS Number: 177908-37-1
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty