Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-Chloro-2-thienyl)methanamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A180707-100MG A180707-1MG A180707-250MG A180707-1G
77151-588EA 13 USD InStock
77151-588 77151-590 77151-592 77685-414
(5-Chloro-2-thienyl)methanamine hydrochloride 97%
(5-Chloro-2-thienyl)methanamine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₇Cl₂NS
MW: 184.09 g/mol
MDL Number: MFCD09971558
CAS Number: 548772-41-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty