Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,3R,4S,5S,6R)-2-(4-((1S,3aR,4R,6aR)-4-(3,4-Dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl)-3-methoxyphenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 95%

Supplier: AMBEED, INC
Warning

A180971-100MG A180971-1MG A180971-250MG A180971-25MG A180971-50MG A180971-1G
77151-594EA 44 USD InStock
77151-594 77151-596 77151-598 77151-600 77151-602 77685-418
(2S,3R,4S,5S,6R)-2-(4-((1S,3aR,4R,6aR)-4-(3,4-Dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl)-3-methoxyphenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 95%
(2S,3R,4S,5S,6R)-2-(4-((1S,3aR,4R,6aR)-4-(3,4-Dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl)-3-methoxyphenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
Precaución: for Research Use Only


Formula: C₂₇H₃₄O₁₁
MDL Number: MFCD00467090
CAS Number: 487-41-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty