Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cobimetinib 98%

Supplier: AMBEED, INC
Warning

A181766-1MG A181766-2MG A181766-5MG A181766-100MG
77151-620EA 17 USD InStock
77151-620 77151-622 77151-624 77685-423
Cobimetinib 98%
Cobimetinib
Precaución: for Research Use Only


Formula: C₂₁H₂₁F₃IN₃O₂
MDL Number: MFCD22124461
CAS Number: 934660-93-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty