Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dibromo-5-fluoro-2-iodobenzene 98%

Supplier: AMBEED, INC
Warning

A196452-100G A196452-1G A196452-25G A196452-5G
77152-024EA 82 USD InStock
77152-024 77152-026 77152-028 77152-030
1,3-Dibromo-5-fluoro-2-iodobenzene 98%
1,3-Dibromo-5-fluoro-2-iodobenzene
Precaución: for Research Use Only


Formula: C₆H₂Br₂FI
MW: 379.79
Boiling Pt: 286
MDL Number: MFCD00042232
CAS Number: 62720-29-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty