Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BAY 80-6946 98%

Supplier: AMBEED, INC
Warning

A199526-10MG A199526-25MG A199526-5MG A199526-50MG
77152-096EA 33 USD InStock
77152-096 77152-098 77152-100 77685-496
BAY 80-6946 98%
BAY 80-6946
Precaución: for Research Use Only


Formula: C₂₃H₂₈N₈O₄
MW: 480.52 g/mol
MDL Number: MFCD18633201
CAS Number: 1032568-63-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty