Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SCH900776 ≥98%

Supplier: AMBEED, INC
Warning

A200833-10MG A200833-25MG A200833-2MG A200833-5MG A200833-100MG A200833-1MG A200833-250MG A200833-50MG
77152-146EA 54 USD InStock
77152-146 77152-148 77152-150 77152-152 77685-505 77685-506 77685-507 77685-508
SCH900776 ≥98%
SCH900776
Precaución: for Research Use Only


Formula: C₁₅H₁₈BrN₇
MDL Number: MFCD20922873
CAS Number: 891494-63-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty