Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6S,9S,12S)-Benzyl 12-benzyl-9-isobutyl-2,2-dimethyl-4,7,10-trioxo-6-phenethyl-3-oxa-5,8,11-triazatridecan-13-oate 95%

Supplier: AMBEED, INC
Warning

A204717-1G A204717-5G
77152-288EA 20 USD InStock
77152-288 77685-521
(6S,9S,12S)-Benzyl 12-benzyl-9-isobutyl-2,2-dimethyl-4,7,10-trioxo-6-phenethyl-3-oxa-5,8,11-triazatridecan-13-oate 95%
(6S,9S,12S)-Benzyl 12-benzyl-9-isobutyl-2,2-dimethyl-4,7,10-trioxo-6-phenethyl-3-oxa-5,8,11-triazatridecan-13-oate
Precaución: for Research Use Only


Formula: C₃₇H₄₇N₃O₆
MDL Number: MFCD29044885
CAS Number: 868540-15-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty