Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzyloxy)-4-bromophenol 95%

Supplier: AMBEED, INC
Warning

A205566-10G A205566-1G A205566-25G A205566-5G
77152-298EA 77 USD InStock
77152-298 77152-300 77152-302 77152-304
2-(Benzyloxy)-4-bromophenol 95%
2-(Benzyloxy)-4-bromophenol
Precaución: for Research Use Only


Formula: C₁₃H₁₁BrO₂
MDL Number: MFCD18433365
CAS Number: 153240-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty