Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-Methyl 2-(2-(bromomethyl)phenyl)-3-methoxyacrylate 95%

Supplier: AMBEED, INC
Danger

A207663-100MG A207663-1G A207663-250MG A207663-5G
77152-348EA 14 USD InStock
77152-348 77152-350 77152-352 77152-354
(E)-Methyl 2-(2-(bromomethyl)phenyl)-3-methoxyacrylate 95%
(E)-Methyl 2-(2-(bromomethyl)phenyl)-3-methoxyacrylate
Precaución: for Research Use Only


Formula: C₁₂H₁₃BrO₃
MDL Number: MFCD16038453
CAS Number: 117428-49-6
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty