Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminophthalic acid hydrochloride dihydrate 95%

Supplier: AMBEED, INC
Warning

A212324-100G A212324-10G A212324-25G A212324-5G
77152-470EA 51 USD InStock
77152-470 77152-472 77152-474 77152-476
3-Aminophthalic acid hydrochloride dihydrate 95%
3-Aminophthalic acid hydrochloride dihydrate
Precaución: for Research Use Only


Formula: C₈H₇NO₄·HCl·2H₂O
MDL Number: MFCD00150282
CAS Number: 6946-22-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty