Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper(I) diphenylphosphinate 90%

Supplier: AMBEED, INC
Warning

A217969-1G A217969-25G A217969-5G
77152-642EA 31 USD InStock
77152-642 77152-644 77152-646
Copper(I) diphenylphosphinate 90%
Copper(I) diphenylphosphinate
Precaución: for Research Use Only


Formula: C₁₂H₁₀CuO₂P
MW: 280.73 g/mol
MDL Number: MFCD09702021
CAS Number: 1011257-42-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty