Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Boc-3-Iodo-4,5,6,7-tetrahydropyrazolo[4,3-c]pyridine 98%

Supplier: AMBEED, INC
Warning

A223275-1G A223275-250MG A223275-5G A223275-10G
77152-736EA 24 USD InStock
77152-736 77152-738 77152-740 77685-601
5-Boc-3-Iodo-4,5,6,7-tetrahydropyrazolo[4,3-c]pyridine 98%
5-Boc-3-Iodo-4,5,6,7-tetrahydropyrazolo[4,3-c]pyridine
Precaución: For research use only.


Formula: C₁₁H₁₆IN₃O₂
MDL Number: MFCD27978466
CAS Number: 661487-17-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty