Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PluriSIn 1 98%

Supplier: AMBEED, INC
Warning

A230785-10MG A230785-1G A230785-1MG A230785-50MG A230785-5MG
77152-876EA 43 USD InStock
77152-876 77152-878 77152-880 77152-882 77152-884
PluriSIn 1 98%
PluriSIn 1
Precaución: for Research Use Only


Formula: C₁₂H₁₁N₃O
MDL Number: MFCD02344904
CAS Number: 91396-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty