Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PF-06463922 98%

Supplier: AMBEED, INC
Warning

A231019-1MG A231019-5MG A231019-250MG
77152-888EA 10 USD InStock
77152-888 77152-890 77568-234
PF-06463922 98%
PF-06463922
Precaución: For research use only.


Formula: C₂₁H₁₉FN₆O₂
MDL Number: MFCD28144520
CAS Number: 1454846-35-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty