Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methylpentanoic acid 95%

Supplier: AMBEED, INC
Warning

A231155-100MG A231155-1G A231155-250MG
77152-892EA 71 USD InStock
77152-892 77152-894 77152-896
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methylpentanoic acid 95%
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methylpentanoic acid
Precaución: for Research Use Only


Formula: C₂₁H₂₃NO₄
MW: 353.41 g/mol
MDL Number: MFCD03095625
CAS Number: 266318-79-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty