Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-carbamoyl-2-methoxybenzoate 95%

Supplier: AMBEED, INC
Warning

A231344-100MG A231344-1G A231344-250MG A231344-50MG
77152-904EA 34 USD InStock
77152-904 77152-906 77152-908 77152-910
Methyl 5-carbamoyl-2-methoxybenzoate 95%
Methyl 5-carbamoyl-2-methoxybenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₄
MDL Number: MFCD24626283
CAS Number: 911281-96-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty