Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

IKK-2 98%

Supplier: AMBEED, INC
Warning

A232738-10MG A232738-1MG A232738-50MG A232738-5MG
77152-928EA 33 USD InStock
77152-928 77152-930 77152-932 77152-934
IKK-2 98%
IKK-2
Precaución: for Research Use Only


Formula: C₁₅H₈ClF₆NO₂
MW: 383.67 g/mol
MDL Number: MFCD00218820
CAS Number: 978-62-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty