Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4',4''-(Benzene-1,3,5-triyltris(oxy))trianiline 95%

Supplier: AMBEED, INC
Warning

A234285-100MG A234285-1G A234285-250MG A234285-5G
77152-960EA 6 USD InStock
77152-960 77152-962 77152-964 77152-966
4,4',4''-(Benzene-1,3,5-triyltris(oxy))trianiline 95%
4,4',4''-(Benzene-1,3,5-triyltris(oxy))trianiline
Precaución: for Research Use Only


Formula: C₂₄H₂₁N₃O₃
MDL Number: MFCD09908237
CAS Number: 102852-92-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty