Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-hydroxy-2-methylbenzoate 97%

Supplier: AMBEED, INC
Warning

A239814-10G A239814-1G A239814-250MG A239814-25G A239814-5G
77153-062EA 20 USD InStock
77153-062 77153-064 77153-066 77153-068 77153-070
Methyl 3-hydroxy-2-methylbenzoate 97%
Methyl 3-hydroxy-2-methylbenzoate
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MW: 166.174 g/mol
MDL Number: MFCD09038292
CAS Number: 55289-05-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty