Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Difluoro-1-boc-azetidine 98+%

Supplier: AMBEED, INC
Warning

A241680-1G A241680-25G A241680-5G
77153-090EA 13 USD InStock
77153-090 77153-092 77153-094
3,3-Difluoro-1-boc-azetidine 98+%
3,3-Difluoro-1-boc-azetidine
Precaución: for Research Use Only


Formula: C₈H₁₃F₂NO₂
MDL Number: MFCD18381087
CAS Number: 1255666-59-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty