Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PYR-41 ≥98%

Supplier: AMBEED, INC
Warning

A242472-100MG A242472-10MG A242472-1MG A242472-25MG A242472-50MG A242472-5MG
77153-114EA 178 USD InStock
77153-114 77153-116 77153-118 77153-120 77153-122 77153-124
PYR-41 ≥98%
PYR-41
Precaución: for Research Use Only


Formula: C₁₇H₁₃N₃O₇·3H₂O
MDL Number: MFCD01469983
CAS Number: 418805-02-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty